Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023630.2 | NP_076119.2 | 6 - 7 | 1005 | 106 | |
GenBank mRNA | AF250835.1 | - | 6 - 7 | 973 | 106 | |
AK005881.1 | - | 6 - 7 | 980 | 106 | ||
BC050756.1 | - | 6 - 7 | 988 | 6741860 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |