Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023668.2 | NP_076157.2 | 4 - 5 | 592 | 86 | |
GenBank mRNA | AF290472.1 | - | 4 - 5 | 524 | 86 | |
AF323918.1 | - | 4 - 5 | 612 | 86 | ||
AK011168.1 | - | 4 - 5 | 502 | 86 | ||
AK146029.1 | - | 4 - 5 | 521 | 86 | ||
BC021434.1 | - | 488 | 5029832 | 86 | ||
BC046796.1 | - | 4 - 5 | 523 | 5718032 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |