Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025351.3 | NP_079627.3 | 4 - 5 | 636 | 63 | |
GenBank mRNA | AK002980.1 | - | 4 - 5 | 620 | 63 | |
AK005696.1 | - | 4 - 5 | 568 | 63 | ||
AK006192.1 | - | 4 - 5 | 572 | 63 | ||
AK015314.1 | - | 4 - 5 | 599 | 63 | ||
AK087642.1 | - | 4 - 5 | 545 | 63 | ||
AK144093.1 | - | 4 - 5 | 542 | 63 | ||
AK167314.1 | - | 4 - 5 | 589 | 63 | ||
BC011331.1 | - | 4 - 5 | 599 | 3153325 | 63 | |
BC052929.1 | - | 4 - 5 | 595 | 1366861 | 63 | |
EF410100.1 | - | 696 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |