Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nicn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025449.3 | NP_079725.1 | 2 - 3 | 379 | 62 | |
GenBank mRNA | AJ299741.1 | - | 2 - 3 | 371 | 62 | |
AK002789.1 | - | 2 - 3 | 385 | 62 | ||
AK013602.1 | - | 2 - 3 | 407 | 62 | ||
AK075732.1 | - | 2 - 3 | 387 | 62 | ||
AK146890.1 | - | 2 - 3 | 389 | 62 | ||
AK169135.1 | - | 2 - 3 | 389 | 62 | ||
BC024050.1 | - | 2 - 3 | 379 | 5133763 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |