Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290706.1 | NP_001277635.1 | 2 - 3 | 379 | 70 | |
NM_025452.4 | NP_079728.2 | 2 - 3 | 397 | 70 | ||
GenBank mRNA | AK008935.1 | - | 2 - 3 | 312 | 70 | |
BC019563.1 | - | 2 - 3 | 326 | 4219997 | 70 | |
BY079646.1 | - | 2 - 3 | 345 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |