Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081274.1 | NP_001074743.1 | 7 - 8 | 765 | 87 | |
GenBank mRNA | AK002894.1 | - | 7 - 8 | 727 | 87 | |
AK036820.1 | - | 1 - 2 | 2037 | 87 | ||
AK145602.1 | - | 7 - 8 | 729 | 87 | ||
AK150210.1 | - | 7 - 8 | 728 | 87 | ||
AK150490.1 | - | 7 - 8 | 724 | 87 | ||
AK151380.1 | - | 7 - 8 | 724 | 87 | ||
AK151993.1 | - | 7 - 8 | 724 | 87 | ||
AK152758.1 | - | 7 - 8 | 724 | 87 | ||
AK152894.1 | - | 7 - 8 | 724 | 87 | ||
AK153409.1 | - | 7 - 8 | 721 | 87 | ||
AK155027.1 | - | 7 - 8 | 724 | 87 | ||
AK161869.1 | - | 6 - 7 | 2686 | 87 | ||
AK166733.1 | - | 7 - 8 | 719 | 87 | ||
AK166947.1 | - | 7 - 8 | 701 | 87 | ||
AK167215.1 | - | 7 - 8 | 719 | 87 | ||
AK168251.1 | - | 7 - 8 | 724 | 87 | ||
BC008646.1 | - | 7 - 8 | 696 | 3155147 | 87 | |
BC011329.1 | - | 7 - 8 | 714 | 3984654 | 87 | |
BC014793.1 | - | 7 - 8 | 733 | 3155100 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |