Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025813.3 | NP_080089.1 | 12 - 13 | 1227 | 102 | |
GenBank mRNA | AK004586.1 | - | 12 - 13 | 1228 | 102 | |
AK032688.1 | - | 12 - 13 | 1195 | 102 | ||
AK075859.1 | - | 12 - 13 | 1257 | 102 | ||
AK135235.1 | - | 12 - 13 | 1228 | 102 | ||
AK152444.1 | - | 12 - 13 | 1249 | 102 | ||
AK159554.1 | - | 12 - 13 | 1255 | 102 | ||
AK166645.1 | - | 6 - 7 | 619 | 102 | ||
AK191801.1 | - | 2 - 3 | 159 | 102 | ||
BC024891.1 | - | 12 - 13 | 1209 | 3992247 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |