Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286560.1 | NP_001273489.1 | 6 - 7 | 809 | 100 | |
NM_026168.4 | NP_080444.1 | 7 - 8 | 918 | 100 | ||
NM_026355.3 | NP_080631.1 | 7 - 8 | 918 | 100 | ||
GenBank mRNA | AK004661.1 | - | 7 - 8 | 582 | 100 | |
AK007498.1 | - | 7 - 8 | 567 | 100 | ||
AK008971.1 | - | 7 - 8 | 574 | 100 | ||
AK009483.1 | - | 7 - 8 | 570 | 100 | ||
AK010529.1 | - | 7 - 8 | 567 | 100 | ||
AK014413.1 | - | 7 - 8 | 574 | 100 | ||
AK016275.1 | - | 7 - 8 | 919 | 100 | ||
AK017876.1 | - | 7 - 8 | 602 | 100 | ||
AK030045.1 | - | 7 - 8 | 568 | 100 | ||
AK086727.1 | - | 7 - 8 | 570 | 100 | ||
AK135968.1 | - | 4 - 5 | 255 | 100 | ||
AK162267.1 | - | 6 - 7 | 542 | 100 | ||
AK167391.1 | - | 7 - 8 | 571 | 100 | ||
BC006895.1 | - | 7 - 8 | 536 | 2651322 | 100 | |
BC018188.1 | - | 7 - 8 | 570 | 3711066 | 100 | |
BC026558.1 | - | 7 - 8 | 530 | 5030661 | 100 | |
BC064749.1 | - | 7 - 8 | 631 | 30046220 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |