Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ict1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026729.1 | NP_081005.1 | 4 - 5 | 286 | 114 | |
GenBank mRNA | AK003192.1 | - | 4 - 5 | 286 | 114 | |
AK003286.1 | - | 4 - 5 | 258 | 114 | ||
BC028523.1 | - | 4 - 5 | 387 | 2938122 | 114 | |
BC079614.1 | - | 325 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |