Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159905.1 | NP_001153377.1 | 2 - 3 | 137 | 93 | |
NM_001159906.1 | NP_001153378.1 | 2 - 3 | 134 | 93 | ||
NM_026846.3 | NP_081122.2 | 1 - 2 | 171 | 93 | ||
GenBank mRNA | AK004332.1 | - | 1 - 2 | 161 | 93 | |
AK151430.1 | - | 1 - 2 | 173 | 93 | ||
BC011495.1 | - | 1 - 2 | 196 | 4196340 | 93 | |
BY197482.1 | - | 2 - 3 | 135 | 93 | ||
CX217579.1 | - | 2 - 3 | 140 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |