Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc46a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027872.3 | NP_082148.1 | 4 - 5 | 1495 | 68 | |
GenBank mRNA | AK004609.1 | - | 4 - 5 | 1280 | 68 | |
AK156898.1 | - | 4 - 5 | 1497 | 68 | ||
BC006902.1 | - | 4 - 5 | 1276 | 3599960 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |