Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027874.2 | NP_082150.1 | 6 - 7 | 1209 | 72 | |
NM_139059.2 | NP_620690.1 | 6 - 7 | 1209 | 72 | ||
GenBank mRNA | AK004606.1 | - | 6 - 7 | 969 | 72 | |
AK051052.1 | - | 6 - 7 | 1208 | 72 | ||
AK088642.1 | - | 6 - 7 | 944 | 72 | ||
AK140317.1 | - | 6 - 7 | 934 | 72 | ||
AK143534.1 | - | 6 - 7 | 910 | 72 | ||
AK146600.1 | - | 6 - 7 | 1199 | 72 | ||
AK150387.1 | - | 6 - 7 | 938 | 72 | ||
AK150815.1 | - | 6 - 7 | 936 | 72 | ||
AK152721.1 | - | 6 - 7 | 1151 | 72 | ||
AK157812.1 | - | 6 - 7 | 944 | 72 | ||
BC004604.1 | - | 6 - 7 | 1195 | 3585286 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |