Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Seh1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039088.1 | NP_001034177.1 | 2 - 3 | 290 | 61 | |
NM_028112.2 | NP_082388.1 | 2 - 3 | 290 | 61 | ||
GenBank mRNA | AK011330.1 | - | 2 - 3 | 192 | 61 | |
AK088835.1 | - | 2 - 3 | 164 | 61 | ||
AK143727.1 | - | 2 - 3 | 290 | 61 | ||
AK166573.1 | - | 2 - 3 | 155 | 61 | ||
AK169231.1 | - | 2 - 3 | 171 | 61 | ||
AK169388.1 | - | 2 - 3 | 170 | 61 | ||
AK170591.1 | - | 2 - 3 | 243 | 61 | ||
BC027244.1 | - | 2 - 3 | 242 | 3584697 | 61 | |
BY732540.1 | - | 2 - 3 | 290 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |