Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028301.4 | NP_082577.2 | 8 - 9 | 1127 | 63 | |
GenBank mRNA | AK012494.1 | - | 8 - 9 | 1076 | 63 | |
AK089037.1 | - | 8 - 9 | 1102 | 63 | ||
AK129471.1 | - | 8 - 9 | 1071 | 63 | ||
BC039644.1 | - | 1 - 2 | 80 | 63 | ||
BC050929.1 | - | 8 - 9 | 1063 | 6807194 | 63 | |
BC051143.1 | - | 8 - 9 | 1094 | 6531161 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |