Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam98c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146023.1 | NP_001139495.1 | 4 - 5 | 574 | 95 | |
GenBank mRNA | AK003496.1 | - | 3 - 4 | 501 | 95 | |
AK080790.1 | - | 3 - 4 | 666 | 95 | ||
AK138395.1 | - | 4 - 5 | 548 | 95 | ||
AK166139.1 | - | 4 - 5 | 544 | 95 | ||
AK171108.1 | - | 2 - 3 | 940 | 95 | ||
BC023421.1 | - | 1 - 2 | 192 | 5029298 | 95 | |
BC058765.1 | - | 3 - 4 | 631 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |