Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028746.3 | NP_083022.1 | 1 - 2 | 765 | 145 | |
GenBank mRNA | AB072352.1 | - | 1 - 2 | 748 | 145 | |
AJ417662.1 | - | 1 - 2 | 714 | 145 | ||
AK128901.1 | - | 1 - 2 | 765 | 145 | ||
AK143768.1 | - | 1 - 2 | 767 | 145 | ||
AK165403.1 | - | 1 - 2 | 766 | 145 | ||
BC014684.1 | - | 1 - 2 | 738 | 4236308 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |