Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB120716.1 | - | 2 - 3 | 185 | 80 | |
AK005610.1 | - | 2 - 3 | 185 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |