Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpe65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029987.2 | NP_084263.2 | 3 - 4 | 289 | 75 | |
GenBank mRNA | AF410461.1 | - | 2 - 3 | 237 | 75 | |
AK020906.1 | - | 3 - 4 | 288 | 75 | ||
BC090656.1 | - | 3 - 4 | 266 | 75 | ||
BC130028.1 | - | 2 - 3 | 241 | 40140883 | 75 | |
CJ133618.1 | - | 3 - 4 | 290 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |