Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpe65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029987.2 | NP_084263.2 | 10 - 11 | 1186 | 103 | |
GenBank mRNA | AF410461.1 | - | 9 - 10 | 1134 | 103 | |
BC090656.1 | - | 10 - 11 | 1162 | 103 | ||
BC130028.1 | - | 9 - 10 | 1138 | 40140883 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |