Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030004.3 | NP_084280.2 | 4 - 5 | 477 | 98 | |
GenBank mRNA | AB359906.1 | - | 4 - 5 | 442 | 98 | |
AF351609.1 | - | 4 - 5 | 522 | 98 | ||
AK003913.1 | - | 4 - 5 | 515 | 98 | ||
AK043569.1 | - | 4 - 5 | 524 | 98 | ||
AK080625.1 | - | 4 - 5 | 498 | 98 | ||
AK135834.1 | - | 4 - 5 | 491 | 98 | ||
AK188247.1 | - | 1 - 2 | 97 | 98 | ||
BC004074.1 | - | 4 - 5 | 477 | 3591359 | 98 | |
BC027064.1 | - | 4 - 5 | 494 | 5355916 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |