Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030251.3 | NP_084527.2 | 6 - 7 | 637 | 81 | |
GenBank mRNA | AB053477.1 | - | 6 - 7 | 568 | 81 | |
AK049100.1 | - | 6 - 7 | 553 | 81 | ||
AK088437.1 | - | 6 - 7 | 602 | 81 | ||
AK089474.1 | - | 4 - 5 | 1001 | 81 | ||
AK220208.1 | - | 6 - 7 | 494 | 81 | ||
BC003234.1 | - | 6 - 7 | 581 | 3483893 | 81 | |
CO804154.1 | - | 3 - 4 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |