Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198914.1 | NP_001185843.1 | 9 - 10 | 1470 | 122 | |
GenBank mRNA | AK088079.1 | - | 9 - 10 | 1391 | 122 | |
CN693253.1 | - | 2 - 3 | 291 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Mus musculus Probe spans exons |