Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dqx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033606.3 | NP_291084.1 | 9 - 10 | 1991 | 62 | |
GenBank mRNA | AF318278.1 | - | 8 - 9 | 1635 | 62 | |
BC048936.1 | - | 4 - 5 | 654 | 62 | ||
BC075680.1 | - | 9 - 10 | 1991 | 30612145 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |