Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Epgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053087.2 | NP_444317.1 | 1 - 2 | 48 | 63 | |
GenBank mRNA | AJ291391.1 | - | 1 - 2 | 48 | 63 | |
BC119291.1 | - | 1 - 2 | 101 | 8734040 | 63 | |
BC119293.1 | - | 1 - 2 | 101 | 8734042 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |