Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130455.2 | NP_569722.1 | 2 - 3 | 1193 | 63 | |
GenBank mRNA | AF396649.1 | - | 2 - 3 | 1202 | 63 | |
AY062435.1 | - | 2 - 3 | 1193 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Mus musculus Probe spans exons |