Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P210603 |
BC003954.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P210603 |
BC058330.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P210603 |
NM_130882.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P210603 |
AF233643.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P210603 |
BC003954.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P210603 |
BC058330.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P210603 |
NM_130882.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P210603 |
AF233643.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932909 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
883535 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
736160 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
828980 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
226170 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
313074 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
162632 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543603 |
AK004443.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543603 |
AK143537.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543603 |
AK147610.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543603 |
AK198397.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543603 |
AK215368.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543603 |
BC014817.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543603 |
NM_028447.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
903962 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
749061 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
411538 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626027 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
603884 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
457989 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
484145 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
396645 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
458640 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
518770 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
36936 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
936534 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
929503 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
10091 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
167721 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
13250 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
52038 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
46847 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
132838 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P112062 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P108781 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P210603 |
BC003954.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P210603 |
BC058330.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P210603 |
NM_130882.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P210603 |
AF233643.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P210603 |
BC003954.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P210603 |
BC058330.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P210603 |
NM_130882.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P210603 |
AF233643.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P210603 |
BC003954.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P210603 |
BC058330.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P210603 |
NM_130882.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P210603 |
AF233643.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P210603 |
BC003954.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P210603 |
BC058330.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P210603 |
NM_130882.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P210603 |
AF233643.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147791 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147783 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147769 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147784 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147771 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147759 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147780 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147767 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147774 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147764 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147773 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147781 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147794 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147770 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147776 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147775 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147761 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147790 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147793 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147796 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147787 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147795 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147792 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147772 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31147778 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08762249 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08762262 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P112062 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08762239 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P108781 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P210603 |
BC003954.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P210603 |
BC058330.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P210603 |
NM_130882.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P210603 |
AF233643.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P210603 |
BC003954.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P210603 |
BC058330.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P210603 |
NM_130882.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P210603 |
AF233643.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P129077 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P08762239 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P108781 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P112062 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13556630 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13556632 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13556623 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13556629 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13556627 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P13556642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147816 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147602 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147699 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31147652 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147781 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147753 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147739 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13556643 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147770 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147805 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147757 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147741 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147737 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147787 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147800 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13556626 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13556629 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147756 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13504515 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147773 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147778 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13158914 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147790 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147796 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13590518 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147744 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147771 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147764 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147746 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13377140 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147802 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147788 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147801 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147784 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147806 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147736 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147758 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147791 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147745 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13556630 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147804 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147779 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147751 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147793 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147799 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147743 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147795 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P13556638 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147747 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147792 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P31147782 |
- |
- |