Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adarb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024837.2 | NP_001020008.1 | 2 - 3 | 350 | 89 | |
NM_130895.3 | NP_570965.2 | 2 - 3 | 350 | 89 | ||
NR_004429.1 | 2 - 3 | 350 | 89 | |||
NR_021486.1 | 2 - 3 | 350 | 89 | |||
GenBank mRNA | AF291049.1 | - | 1 - 2 | 185 | 89 | |
AF403106.2 | - | 2 - 3 | 302 | 89 | ||
AF403107.2 | - | 2 - 3 | 302 | 89 | ||
AF403108.2 | - | 2 - 3 | 302 | 89 | ||
AF403109.2 | - | 2 - 3 | 302 | 89 | ||
AF525421.1 | - | 2 - 3 | 302 | 89 | ||
AK006823.1 | - | 3 - 4 | 302 | 89 | ||
AK138318.1 | - | 2 - 3 | 343 | 89 | ||
AK141777.1 | - | 2 - 3 | 322 | 89 | ||
AK147298.1 | - | 3 - 4 | 429 | 89 | ||
AK164931.1 | - | 2 - 3 | 318 | 89 | ||
AY162454.1 | - | 2 - 3 | 302 | 89 | ||
BB622419.1 | - | 2 - 3 | 350 | 89 | ||
BC051107.1 | - | 2 - 3 | 288 | 89 | ||
BC062806.1 | - | 2 - 3 | 305 | 89 | ||
BF017513.1 | - | 1 - 2 | 89 | |||
CJ115965.1 | - | 2 - 3 | 350 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |