Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edaradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133643.4 | NP_598398.3 | 2 - 3 | 598 | 60 | |
GenBank mRNA | AF358671.1 | - | 2 - 3 | 592 | 60 | |
AK041545.1 | - | 2 - 3 | 532 | 60 | ||
AK042128.1 | - | 2 - 3 | 256 | 60 | ||
AK042414.1 | - | 2 - 3 | 561 | 60 | ||
AK075786.1 | - | 2 - 3 | 234 | 60 | ||
AK148351.1 | - | 2 - 3 | 420 | 60 | ||
AY071863.1 | - | 2 - 3 | 96 | 60 | ||
BC030683.1 | - | 2 - 3 | 174 | 1344937 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |