Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB073078.1 | - | 5 - 6 | 761 | 78 | |
AB073079.1 | - | 5 - 6 | 762 | 78 | ||
AB073080.1 | - | 5 - 6 | 761 | 78 | ||
AK050922.1 | - | 6 - 7 | 862 | 78 | ||
AK078113.1 | - | 4 - 5 | 653 | 78 | ||
AK142379.1 | - | 4 - 5 | 672 | 78 | ||
BC145175.1 | - | 4 - 5 | 652 | 9053726 | 78 | |
BC145176.1 | - | 4 - 5 | 652 | 9053727 | 78 | |
BC145850.1 | - | 4 - 5 | 652 | 40131153 | 78 | |
S66169.1 | - | 1 - 2 | 74 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |