Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133664.3 | NP_598425.2 | 6 - 7 | 1661 | 81 | |
GenBank mRNA | AK083526.1 | - | 6 - 7 | 1662 | 81 | |
BC016257.1 | - | 6 - 7 | 1332 | 4912735 | 81 | |
BC025580.1 | - | 6 - 7 | 1565 | 5325039 | 81 | |
BC031131.1 | - | 6 - 7 | 1508 | 4954891 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |