Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133735.2 | NP_598496.3 | 4 - 5 | 980 | 90 | |
GenBank mRNA | AK088764.1 | - | 4 - 5 | 899 | 90 | |
AK143877.1 | - | 4 - 5 | 937 | 90 | ||
AK150514.1 | - | 4 - 5 | 937 | 90 | ||
AK151064.1 | - | 4 - 5 | 948 | 90 | ||
AK152077.1 | - | 4 - 5 | 948 | 90 | ||
AK153366.1 | - | 4 - 5 | 948 | 90 | ||
AK171484.1 | - | 4 - 5 | 935 | 90 | ||
BC004766.1 | - | 4 - 5 | 898 | 3156304 | 90 | |
BC019121.1 | - | 2 - 3 | 368 | 90 | ||
BC083097.1 | - | 4 - 5 | 939 | 30459747 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |