Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133756.4 | NP_598517.1 | 4 - 5 | 325 | 66 | |
GenBank mRNA | AK010393.1 | - | 4 - 5 | 339 | 66 | |
AK077645.1 | - | 4 - 5 | 326 | 66 | ||
AK145957.1 | - | 322 | 66 | |||
AK146686.1 | - | 337 | 66 | |||
BC020174.1 | - | 4 - 5 | 325 | 4505118 | 66 | |
BC096466.1 | - | 4 - 5 | 318 | 3711417 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |