Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133765.4 | NP_598526.3 | 1 - 2 | 349 | 65 | |
GenBank mRNA | AB041586.1 | - | 1 - 2 | 198 | 65 | |
AK140412.1 | - | 1 - 2 | 350 | 65 | ||
AK163641.1 | - | 1 - 2 | 318 | 65 | ||
BC026929.1 | - | 1 - 2 | 334 | 4194857 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |