Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282118.1 | NP_001269047.1 | 4 - 5 | 677 | 132 | |
NM_001282119.1 | NP_001269048.1 | 4 - 5 | 599 | 132 | ||
NM_001282120.1 | NP_001269049.1 | 4 - 5 | 1041 | 132 | ||
NM_133767.3 | NP_598528.2 | 4 - 5 | 955 | 132 | ||
GenBank mRNA | AK010766.1 | - | 4 - 5 | 900 | 132 | |
AK136344.1 | - | 4 - 5 | 639 | 132 | ||
AK146961.1 | - | 4 - 5 | 895 | 132 | ||
BC016590.1 | - | 4 - 5 | 531 | 3585359 | 132 | |
BC066819.1 | - | 4 - 5 | 935 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |