Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437267 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437263 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437268 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437266 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437269 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437265 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437264 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P144926 |
BC021488.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P144926 |
BC024421.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P144926 |
AK187619.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P144926 |
BC017690.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P144926 |
BC021488.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P144926 |
BC024421.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P144926 |
AK187619.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P144926 |
BC017690.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
95962 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
140131 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
788159 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693389 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776650 |
AK154786.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776650 |
AK170037.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776650 |
AY046550.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776650 |
AY165625.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776650 |
BC106828.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776650 |
BC106829.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776650 |
NM_024290.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
176258 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
155157 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P116077 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P138463 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P144926 |
BC021488.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P144926 |
BC024421.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P144926 |
AK187619.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P144926 |
BC017690.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P144926 |
BC021488.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P144926 |
BC024421.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P144926 |
AK187619.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P144926 |
BC017690.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437230 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437272 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437254 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437269 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437264 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437252 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437250 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437229 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437239 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437280 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437281 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437273 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437248 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437237 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437260 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437275 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437277 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437268 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437255 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437234 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437244 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437282 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437242 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437258 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437278 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437233 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437263 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437274 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437231 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437266 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437240 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437246 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20437262 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P138463 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04185561 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P116077 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P144926 |
BC021488.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P144926 |
BC024421.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P144926 |
AK187619.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P144926 |
BC017690.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P144926 |
BC021488.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P144926 |
BC024421.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P144926 |
AK187619.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P144926 |
BC017690.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P128117 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P138463 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P116077 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00843023 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00131184 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00131179 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00071811 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00071812 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00843024 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00131182 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437263 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437264 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437266 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437267 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437265 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437269 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P20437268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437326 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437325 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437263 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437364 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437370 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437328 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437384 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437279 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437339 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437228 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437274 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437273 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437287 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437289 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437280 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437390 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437360 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437386 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437290 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437307 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437266 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437268 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437308 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437373 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437240 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437318 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437304 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437346 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437313 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437341 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437221 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437321 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437299 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437244 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437276 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437219 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437324 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437393 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437322 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437357 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437297 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437227 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437248 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437258 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437342 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437334 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437348 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437332 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437305 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437247 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437344 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437294 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437292 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437277 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437281 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437362 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437366 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437260 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437225 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437316 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437353 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20437301 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437290 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437289 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437281 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00131179 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437318 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00317887 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00341737 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00556728 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437297 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00159055 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437310 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437292 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00697613 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437299 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437259 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00131182 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00341738 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00156876 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437237 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00365369 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437257 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437250 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00922468 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437255 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437276 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437235 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437272 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00375280 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437287 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437322 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437242 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437305 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437301 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437320 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437313 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437303 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437225 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437227 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437308 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437260 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437286 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00365366 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00375279 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01005557 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437288 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437285 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437273 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437279 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00131184 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00071812 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00557759 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00843024 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437316 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00557762 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437296 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437283 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437239 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437269 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437234 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P00365364 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437247 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437252 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437324 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20437277 |
- |
- |