Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133838.4 | NP_598599.2 | 2 - 3 | 506 | 100 | |
GenBank mRNA | AB239915.1 | - | 2 - 3 | 417 | 100 | |
AB239916.1 | - | 2 - 3 | 417 | 100 | ||
AF173639.1 | - | 2 - 3 | 422 | 100 | ||
AK147186.1 | - | 2 - 3 | 458 | 100 | ||
AK149760.1 | - | 2 - 3 | 450 | 100 | ||
AK159235.1 | - | 2 - 3 | 466 | 100 | ||
AK159597.1 | - | 2 - 3 | 466 | 100 | ||
AK163835.1 | - | 2 - 3 | 450 | 100 | ||
AK166101.1 | - | 2 - 3 | 517 | 100 | ||
AK169343.1 | - | 2 - 3 | 517 | 100 | ||
BC007480.1 | - | 2 - 3 | 546 | 3493227 | 100 | |
BC012272.1 | - | 2 - 3 | 439 | 3485836 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |