Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042652.1 | NP_001036117.1 | 5 - 6 | 1096 | 100 | |
NM_133851.3 | NP_598612.1 | 6 - 7 | 1195 | 100 | ||
GenBank mRNA | AF305710.1 | - | 6 - 7 | 776 | 100 | |
AK042697.1 | - | 677 | 100 | |||
AK044819.1 | - | 6 - 7 | 768 | 100 | ||
AK076028.1 | - | 6 - 7 | 765 | 100 | ||
AK088389.1 | - | 6 - 7 | 795 | 100 | ||
BC004787.1 | - | 6 - 7 | 745 | 3588697 | 100 | |
BC009096.1 | - | 351 | 100 | |||
BC100392.1 | - | 6 - 7 | 1195 | 30110191 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |