Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080968.1 | NP_001074437.1 | 24 - 25 | 2691 | 76 | |
NM_133852.2 | NP_598613.2 | 25 - 26 | 2772 | 76 | ||
GenBank mRNA | AK076370.1 | - | 19 - 20 | 2390 | 76 | |
AK077516.1 | - | 23 - 24 | 2713 | 76 | ||
AK220509.1 | - | 25 - 26 | 2700 | 76 | ||
BC011407.1 | - | 23 - 24 | 2492 | 3596079 | 76 | |
BC024099.1 | - | 1 - 2 | 97 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |