Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133879.2 | NP_598640.2 | 5 - 6 | 1249 | 95 | |
GenBank mRNA | AK027912.1 | - | 4 - 5 | 1733 | 95 | |
AM161454.1 | - | 5 - 6 | 1197 | 95 | ||
BC005515.1 | - | 5 - 6 | 1260 | 3493611 | 95 | |
CX202255.1 | - | 3 - 4 | 426 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |