Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290742.1 | NP_001277671.1 | 3 - 4 | 741 | 96 | |
NM_133884.2 | NP_598645.2 | 3 - 4 | 810 | 96 | ||
GenBank mRNA | BC018407.1 | - | 3 - 4 | 803 | 4015340 | 96 |
CN721622.1 | - | 3 - 4 | 412 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |