Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpdl3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133888.2 | NP_598649.1 | 6 - 7 | 996 | 65 | |
GenBank mRNA | AK136602.1 | - | 5 - 6 | 658 | 65 | |
AK154599.1 | - | 6 - 7 | 1022 | 65 | ||
AK166253.1 | - | 6 - 7 | 972 | 65 | ||
BC009087.1 | - | 6 - 7 | 996 | 2810972 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |