Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133939.1 | NP_598700.1 | 2 - 3 | 172 | 100 | |
GenBank mRNA | AK008067.1 | - | 2 - 3 | 154 | 100 | |
AK088664.1 | - | 2 - 3 | 133 | 100 | ||
AK154446.1 | - | 1037 | 100 | |||
AK167674.1 | - | 104 | 100 | |||
BC019458.1 | - | 2 - 3 | 172 | 4024697 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |