Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133952.2 | NP_598713.2 | 8 - 9 | 1120 | 63 | |
GenBank mRNA | AK028912.1 | - | 8 - 9 | 1106 | 63 | |
AK047292.1 | - | 8 - 9 | 1067 | 63 | ||
AK047476.1 | - | 8 - 9 | 1067 | 63 | ||
AK088034.1 | - | 8 - 9 | 1072 | 63 | ||
AK166809.1 | - | 8 - 9 | 1103 | 63 | ||
BC004717.1 | - | 8 - 9 | 1109 | 3582116 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |