Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142809.1 | NP_001136281.1 | 7 - 8 | 1799 | 65 | |
NM_001142810.1 | NP_001136282.1 | 7 - 8 | 1799 | 65 | ||
NM_133987.2 | NP_598748.1 | 7 - 8 | 1799 | 65 | ||
GenBank mRNA | AB077327.1 | - | 7 - 8 | 1147 | 65 | |
AF459435.1 | - | 7 - 8 | 1782 | 65 | ||
AJ748650.1 | - | 4 - 5 | 65 | |||
AK189504.1 | - | 4 - 5 | 219 | 65 | ||
BC024444.1 | - | 6 - 7 | 812 | 65 | ||
BC029000.1 | - | 5 - 6 | 562 | 65 | ||
BC049801.1 | - | 7 - 8 | 974 | 65 | ||
BC141067.1 | - | 8 - 9 | 1540 | 9055679 | 65 | |
BC145319.1 | - | 8 - 9 | 1204 | 9053874 | 65 | |
EU523148.1 | - | 3 - 4 | 848 | 65 | ||
KC994641.1 | - | 3 - 4 | 848 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |