Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159591.1 | NP_001153063.1 | 6 - 7 | 997 | 66 | |
NM_134002.2 | NP_598763.1 | 5 - 6 | 1174 | 66 | ||
GenBank mRNA | AK077076.1 | - | 6 - 7 | 977 | 66 | |
AK132871.1 | - | 6 - 7 | 998 | 66 | ||
AK152413.1 | - | 5 - 6 | 1173 | 66 | ||
BC004839.1 | - | 5 - 6 | 1116 | 2648652 | 66 | |
BC063083.1 | - | 3 - 4 | 2298 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |