Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134034.2 | NP_598795.2 | 1 - 2 | 547 | 84 | |
GenBank mRNA | AK037253.1 | - | 1 - 2 | 479 | 84 | |
AK041513.1 | - | 1 - 2 | 492 | 84 | ||
AK044742.1 | - | 1 - 2 | 488 | 84 | ||
AK090014.1 | - | 1 - 2 | 523 | 84 | ||
AK129344.1 | - | 1 - 2 | 458 | 84 | ||
AK165940.1 | - | 1 - 2 | 471 | 84 | ||
AK166627.1 | - | 1 - 2 | 473 | 84 | ||
BC006870.1 | - | 1 - 2 | 458 | 3595439 | 84 | |
CD350450.1 | - | 1 - 2 | 547 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |