Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134093.2 | NP_598854.1 | 6 - 7 | 851 | 69 | |
GenBank mRNA | AF287293.1 | - | 6 - 7 | 791 | 69 | |
AF401483.1 | - | 6 - 7 | 684 | 69 | ||
AK041312.1 | - | 5 - 6 | 1180 | 69 | ||
AK045256.1 | - | 6 - 7 | 671 | 69 | ||
AK046496.1 | - | 4 - 5 | 751 | 69 | ||
AK080221.1 | - | 6 - 7 | 771 | 69 | ||
AK154676.1 | - | 6 - 7 | 766 | 69 | ||
AK158718.1 | - | 5 - 6 | 628 | 69 | ||
AK169341.1 | - | 6 - 7 | 771 | 69 | ||
BC021361.1 | - | 3 - 4 | 523 | 5010892 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |