Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134127.1 | NP_598888.1 | 4 - 5 | 551 | 81 | |
GenBank mRNA | AF233645.1 | - | 4 - 5 | 551 | 81 | |
BC021377.1 | - | 4 - 5 | 542 | 5050617 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |