Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083902.1 | NP_001077371.1 | 2 - 3 | 224 | 83 | |
NM_134251.2 | NP_599012.2 | 2 - 3 | 193 | 83 | ||
GenBank mRNA | AF319951.1 | - | 2 - 3 | 187 | 83 | |
BC030926.1 | - | 2 - 3 | 224 | 4910321 | 83 | |
BU921470.1 | - | 3 - 4 | 410 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |