Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134420.4 | NP_599252.2 | 2570 | 65 | ||
GenBank mRNA | AB099881.1 | - | 1731 | 65 | ||
AF248494.1 | - | 1916 | 65 | |||
AK029238.1 | - | 3492 | 65 | |||
AK046993.1 | - | 3830 | 65 | |||
AK053088.1 | - | 2221 | 65 | |||
AK163670.1 | - | 2571 | 65 | |||
AK163671.1 | - | 2571 | 65 | |||
AY032863.1 | - | 1875 | 65 | |||
AY049076.1 | - | 1830 | 65 | |||
BC028856.1 | - | 1894 | 4165725 | 65 | ||
CN692543.1 | - | 390 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |